Assay Details
Gene Symbol: LOC100146423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283088.1 | NP_001270017.1 | 28 - 29 | 3314 | 76 | |
XM_023621990.1 | XP_023477758.1 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
