Assay Details
Gene Symbol: MMADHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286855.1 | NP_001273784.1 | 5 - 6 | 585 | 64 | |
GenBank mRNA | CX605249.1 | - | 2 - 3 | 142 | 64 | |
HQ889879.1 | - | 5 - 6 | 585 | 64 | ||
HQ890283.1 | - | 5 - 6 | 566 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
