Assay Details
Gene Symbol: TNFSF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297473.1 | NP_001284402.1 | 4 - 5 | 418 | 68 | |
GenBank mRNA | JL620193.1 | - | 1 - 2 | 1161 | 68 | |
KJ584317.1 | - | 4 - 5 | 418 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
