Assay Details
Gene Symbol: CERS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301183.2 | NP_001288112.1 | 4 - 5 | 607 | 61 | |
XM_001497253.5 | XP_001497303.1 | 652 | 61 | |||
XM_014732229.2 | XP_014587715.1 | 1502 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
