Assay Details
Gene Symbol: SAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301290.1 | NP_001288219.1 | 3 - 4 | 323 | 60 | |
GenBank mRNA | BI961849.1 | - | 1 - 2 | 114 | 60 | |
CD470832.1 | - | 3 - 4 | 323 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
