Assay Details
Gene Symbol: EIF3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301291.1 | NP_001288220.1 | 12 - 13 | 1327 | 81 | |
GenBank mRNA | HQ889978.1 | - | 8 - 9 | 926 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
