Assay Details
Gene Symbol: APC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301314.1 | NP_001288243.1 | 11 - 12 | 1447 | 88 | |
XM_005599506.3 | XP_005599563.1 | 1552 | 88 | |||
XM_005599508.3 | XP_005599565.1 | 1249 | 88 | |||
XM_014730328.2 | XP_014585814.1 | 1440 | 88 | |||
XM_014730331.2 | XP_014585817.1 | 1137 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
