Assay Details
Gene Symbol: APC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301314.1 | NP_001288243.1 | 14 - 15 | 1786 | 76 | |
XM_005599506.3 | XP_005599563.1 | 1891 | 76 | |||
XM_005599508.3 | XP_005599565.1 | 1588 | 76 | |||
XM_014730325.2 | XP_014585811.1 | 1945 | 76 | |||
XM_014730326.2 | XP_014585812.1 | 1908 | 76 | |||
XM_014730327.2 | XP_014585813.1 | 1967 | 76 | |||
XM_014730328.2 | XP_014585814.1 | 1779 | 76 | |||
XM_014730329.2 | XP_014585815.1 | 1987 | 76 | |||
XM_014730330.2 | XP_014585816.1 | 1684 | 76 | |||
XM_014730331.2 | XP_014585817.1 | 1476 | 76 | |||
XM_023616923.1 | XP_023472691.1 | 1934 | 76 | |||
XM_023616924.1 | XP_023472692.1 | 1846 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
