Assay Details
Gene Symbol: APC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301314.1 | NP_001288243.1 | 3 - 4 | 252 | 77 | |
XM_005599506.3 | XP_005599563.1 | 357 | 77 | |||
XM_005599508.3 | XP_005599565.1 | 357 | 77 | |||
XM_014730325.2 | XP_014585811.1 | 357 | 77 | |||
XM_014730326.2 | XP_014585812.1 | 320 | 77 | |||
XM_014730327.2 | XP_014585813.1 | 379 | 77 | |||
XM_014730328.2 | XP_014585814.1 | 245 | 77 | |||
XM_014730329.2 | XP_014585815.1 | 399 | 77 | |||
XM_014730330.2 | XP_014585816.1 | 399 | 77 | |||
XM_014730331.2 | XP_014585817.1 | 245 | 77 | |||
XM_023616923.1 | XP_023472691.1 | 346 | 77 | |||
XM_023616924.1 | XP_023472692.1 | 258 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
