Assay Details
Gene Symbol: APC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301314.1 | NP_001288243.1 | 5 - 6 | 569 | 119 | |
XM_005599506.3 | XP_005599563.1 | 674 | 119 | |||
XM_005599508.3 | XP_005599565.1 | 674 | 119 | |||
XM_014730325.2 | XP_014585811.1 | 674 | 119 | |||
XM_014730326.2 | XP_014585812.1 | 637 | 119 | |||
XM_014730327.2 | XP_014585813.1 | 696 | 119 | |||
XM_014730328.2 | XP_014585814.1 | 562 | 119 | |||
XM_014730329.2 | XP_014585815.1 | 716 | 119 | |||
XM_014730330.2 | XP_014585816.1 | 716 | 119 | |||
XM_014730331.2 | XP_014585817.1 | 562 | 119 | |||
XM_023616923.1 | XP_023472691.1 | 663 | 119 | |||
XM_023616924.1 | XP_023472692.1 | 575 | 119 | |||
GenBank mRNA | JL629200.1 | - | 1 - 2 | 67 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
