Assay Details
Gene Symbol: CAB39L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309165.1 | NP_001296094.1 | 4 - 5 | 588 | 84 | |
XM_001489141.5 | XP_001489191.1 | 1096 | 84 | |||
XM_005601212.3 | XP_005601269.1 | 895 | 84 | |||
XM_005601213.3 | XP_005601270.1 | 1038 | 84 | |||
XM_005601215.3 | XP_005601272.1 | 892 | 84 | |||
XM_014732021.2 | XP_014587507.1 | 1146 | 84 | |||
XM_023621308.1 | XP_023477076.1 | 1138 | 84 | |||
XM_023621309.1 | XP_023477077.1 | 1263 | 84 | |||
XM_023621310.1 | XP_023477078.1 | 1260 | 84 | |||
XM_023621311.1 | XP_023477079.1 | 1035 | 84 | |||
GenBank mRNA | JL616045.1 | - | 4 - 5 | 582 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
