Assay Details
Gene Symbol: NRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309170.2 | NP_001296099.1 | 1 - 2 | 111 | 89 | |
GenBank mRNA | JL615886.1 | - | 1 - 2 | 111 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001309170.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
