Assay Details
Gene Symbol: NRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309170.2 | NP_001296099.1 | 2 - 3 | 232 | 98 | |
GenBank mRNA | JL615886.1 | - | 2 - 3 | 232 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
