Assay Details
Gene Symbol: DNTTIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309173.2 | NP_001296102.1 | 5 - 6 | 2049 | 66 | |
GenBank mRNA | CX596239.1 | - | 3 - 4 | 286 | 66 | |
CX603897.1 | - | 1 - 2 | 186 | 66 | ||
HQ890286.1 | - | 4 - 5 | 887 | 66 | ||
JL637892.1 | - | 2 - 3 | 268 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
