Assay Details
Gene Symbol: DNTTIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309173.2 | NP_001296102.1 | 6 - 7 | 2154 | 68 | |
GenBank mRNA | CX596239.1 | - | 4 - 5 | 391 | 68 | |
CX603897.1 | - | 2 - 3 | 291 | 68 | ||
JL637892.1 | - | 3 - 4 | 373 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
