Assay Details
Gene Symbol: NUP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309191.1 | NP_001296120.1 | 2 - 3 | 265 | 70 | |
XM_014732230.2 | XP_014587716.1 | 291 | 70 | |||
XM_023621971.1 | XP_023477739.1 | 291 | 70 | |||
GenBank mRNA | JL616048.2 | - | 2 - 3 | 265 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
