Assay Details
Gene Symbol: RFC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309197.1 | NP_001296126.1 | 7 - 8 | 669 | 61 | |
XM_023646588.1 | XP_023502356.1 | 675 | 61 | |||
GenBank mRNA | HQ890062.1 | - | 4 - 5 | 384 | 61 | |
JL618054.1 | - | 5 - 6 | 477 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
