Assay Details
Gene Symbol: TBPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309243.1 | NP_001296172.1 | 3 - 4 | 282 | 68 | |
XM_023649386.1 | XP_023505154.1 | 674 | 68 | |||
XM_023649388.1 | XP_023505156.1 | 626 | 68 | |||
XM_023649389.1 | XP_023505157.1 | 471 | 68 | |||
XM_023649390.1 | XP_023505158.1 | 467 | 68 | |||
XM_023649391.1 | XP_023505159.1 | 439 | 68 | |||
XM_023649392.1 | XP_023505160.1 | 485 | 68 | |||
GenBank mRNA | JL616467.1 | - | 3 - 4 | 283 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
