Assay Details
Gene Symbol: RPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309246.1 | NP_001296175.1 | 7 - 8 | 492 | 99 | |
XM_023649400.1 | XP_023505168.1 | 576 | 99 | |||
XM_023649402.1 | XP_023505170.1 | 648 | 99 | |||
GenBank mRNA | JL619273.1 | - | 7 - 8 | 492 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
