Assay Details
Gene Symbol: EIF3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309259.1 | NP_001296188.1 | 3 - 4 | 451 | 70 | |
XM_023648312.1 | XP_023504080.1 | 641 | 70 | |||
GenBank mRNA | JL616206.1 | - | 3 - 4 | 451 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
