Assay Details
Gene Symbol: XRCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309266.1 | NP_001296195.1 | 2 - 3 | 167 | 114 | |
GenBank mRNA | JL616277.2 | - | 2 - 3 | 167 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
