Assay Details
Gene Symbol: DCAF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309279.1 | NP_001296208.1 | 10 - 11 | 1329 | 64 | |
GenBank mRNA | HQ890045.1 | - | 7 - 8 | 817 | 64 | |
JL616540.2 | - | 10 - 11 | 1343 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
