Assay Details
Gene Symbol: COPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309280.1 | NP_001296209.1 | 6 - 7 | 928 | 67 | |
XM_005613085.3 | XP_005613142.1 | 926 | 67 | |||
GenBank mRNA | JL616410.2 | - | 6 - 7 | 920 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
