Assay Details
Gene Symbol: ATP6V1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309283.1 | NP_001296212.1 | 1 - 2 | 86 | 80 |
Gene Symbol: ATP6V1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023648795.1 | XP_023504563.1 | 339 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001309283.1, XM_023648795.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
