Assay Details
Gene Symbol: ATP6V1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309283.1 | NP_001296212.1 | 5 - 6 | 500 | 85 |
Gene Symbol: ATP6V1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023648793.1 | XP_023504561.1 | 570 | 85 | ||
XM_023648795.1 | XP_023504563.1 | 753 | 85 | |||
XM_023648796.1 | XP_023504564.1 | 572 | 85 | |||
GenBank mRNA | JL617032.1 | - | 5 - 6 | 546 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
