Assay Details
Gene Symbol: ATP6V1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309283.1 | NP_001296212.1 | 8 - 9 | 755 | 94 |
Gene Symbol: ATP6V1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023648793.1 | XP_023504561.1 | 825 | 94 | ||
XM_023648795.1 | XP_023504563.1 | 1008 | 94 | |||
GenBank mRNA | JL617032.1 | - | 8 - 9 | 801 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
