Assay Details
Gene Symbol: BLVRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309290.1 | NP_001296219.1 | 4 - 5 | 366 | 60 | |
XM_005609106.3 | XP_005609163.1 | 402 | 60 | |||
XM_005609107.3 | XP_005609164.1 | 424 | 60 | |||
XM_014738889.2 | XP_014594375.2 | 970 | 60 | |||
XM_023638772.1 | XP_023494540.1 | 834 | 60 | |||
XM_023638773.1 | XP_023494541.1 | 804 | 60 | |||
XM_023638774.1 | XP_023494542.1 | 416 | 60 | |||
GenBank mRNA | JL616290.1 | - | 4 - 5 | 392 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
