Assay Details
Gene Symbol: SSR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309293.1 | NP_001296222.1 | 2 - 3 | 291 | 101 | |
GenBank mRNA | DN506945.1 | - | 2 - 3 | 291 | 101 | |
JL624136.1 | - | 2 - 3 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
