Assay Details
Gene Symbol: SSR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309293.1 | NP_001296222.1 | 4 - 5 | 521 | 86 | |
GenBank mRNA | BM734771.1 | - | 2 - 3 | 197 | 86 | |
DN506945.1 | - | 4 - 5 | 521 | 86 | ||
JL624136.1 | - | 4 - 5 | 387 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
