Assay Details
Gene Symbol: MMUT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309313.2 | NP_001296242.1 | 10 - 11 | 1990 | 76 | |
XM_001502799.5 | XP_001502849.1 | 2088 | 76 | |||
XM_014734210.2 | XP_014589696.1 | 2133 | 76 | |||
XM_014734211.2 | XP_014589697.1 | 2097 | 76 | |||
GenBank mRNA | HQ890001.1 | - | 5 - 6 | 76 | ||
JL640521.1 | - | 7 - 8 | 893 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
