Assay Details
Gene Symbol: MMUT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309313.2 | NP_001296242.1 | 3 - 4 | 956 | 67 | |
XM_001502799.5 | XP_001502849.1 | 1054 | 67 | |||
XM_014734210.2 | XP_014589696.1 | 1099 | 67 | |||
XM_014734211.2 | XP_014589697.1 | 1063 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
