Assay Details
Gene Symbol: MMUT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309313.2 | NP_001296242.1 | 6 - 7 | 1482 | 102 | |
XM_001502799.5 | XP_001502849.1 | 1580 | 102 | |||
XM_014734210.2 | XP_014589696.1 | 1625 | 102 | |||
XM_014734211.2 | XP_014589697.1 | 1589 | 102 | |||
GenBank mRNA | HQ890001.1 | - | 1 - 2 | 56 | 102 | |
JL640521.1 | - | 3 - 4 | 385 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
