Assay Details
Gene Symbol: ARL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309322.1 | NP_001296251.1 | 3 - 4 | 295 | 98 | |
XM_014736715.2 | XP_014592201.1 | 266 | 98 | |||
GenBank mRNA | CX605286.1 | - | 3 - 4 | 318 | 98 | |
JL615880.1 | - | 2 - 3 | 243 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
