Assay Details
Gene Symbol: FDPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309326.1 | NP_001296255.1 | 9 - 10 | 1047 | 57 | |
XM_003365005.4 | XP_003365053.1 | 1268 | 57 | |||
XM_005610039.3 | XP_005610096.1 | 1029 | 57 | |||
GenBank mRNA | CD466912.1 | - | 4 - 5 | 340 | 57 | |
JL615944.1 | - | 9 - 10 | 1050 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
