Assay Details
Gene Symbol: SSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309328.1 | NP_001296257.1 | 3 - 4 | 303 | 64 | |
XM_014739706.1 | XP_014595192.1 | 382 | 64 | |||
GenBank mRNA | CD470449.1 | - | 3 - 4 | 303 | 64 | |
CX592675.1 | - | 3 - 4 | 277 | 64 | ||
JL616028.1 | - | 3 - 4 | 298 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
