Assay Details
Gene Symbol: SNUPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309333.1 | NP_001296262.1 | 2 - 3 | 186 | 78 | |
XM_023635726.1 | XP_023491494.1 | 288 | 78 | |||
XM_023635785.1 | XP_023491553.1 | 298 | 78 | |||
GenBank mRNA | JL615911.1 | - | 2 - 3 | 186 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
