Assay Details
Gene Symbol: CASP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309343.2 | NP_001296272.1 | 4 - 5 | 584 | 71 | |
GenBank mRNA | JL616394.1 | - | 4 - 5 | 584 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
