Assay Details
Gene Symbol: PPP3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309345.1 | NP_001296274.1 | 1 - 2 | 40 | 79 | |
XM_023618419.1 | XP_023474187.1 | 153 | 79 | |||
XM_023618420.1 | XP_023474188.1 | 189 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001309345.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
