Assay Details
Gene Symbol: PPP3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309345.1 | NP_001296274.1 | 2 - 3 | 219 | 94 | |
XM_023618419.1 | XP_023474187.1 | 332 | 94 | |||
XM_023618420.1 | XP_023474188.1 | 368 | 94 | |||
GenBank mRNA | JL629851.1 | - | 2 - 3 | 673 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
