Assay Details
Gene Symbol: PPP3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309345.1 | NP_001296274.1 | 4 - 5 | 453 | 98 | |
XM_023618419.1 | XP_023474187.1 | 566 | 98 | |||
XM_023618420.1 | XP_023474188.1 | 602 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
