Assay Details
Gene Symbol: NMNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309407.1 | NP_001296336.1 | 1 - 2 | 169 | 73 | |
XM_005607545.3 | XP_005607602.1 | 281 | 73 | |||
GenBank mRNA | JL616281.2 | - | 1 - 2 | 177 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
