Assay Details
Gene Symbol: NMNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309407.1 | NP_001296336.1 | 2 - 3 | 350 | 84 | |
XM_005607545.3 | XP_005607602.1 | 462 | 84 | |||
GenBank mRNA | JL616281.2 | - | 2 - 3 | 358 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
