Assay Details
Gene Symbol: RAB2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309415.1 | NP_001296344.1 | 4 - 5 | 285 | 56 | |
GenBank mRNA | JL616272.2 | - | 4 - 5 | 288 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
