Assay Details
Gene Symbol: RPS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309481.1 | NP_001296410.1 | 2 - 3 | 95 | 94 | |
GenBank mRNA | CD466713.1 | - | 2 - 3 | 94 | 94 | |
CX593239.1 | - | 2 - 3 | 118 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001309481.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
