Assay Details
Gene Symbol: SLCO1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001317251.1 | NP_001304180.1 | 11 - 12 | 1714 | 66 | |
XM_005610968.3 | XP_005611025.1 | 2143 | 66 | |||
XM_023642200.1 | XP_023497968.1 | 2148 | 66 | |||
XM_023642201.1 | XP_023497969.1 | 2005 | 66 | |||
GenBank mRNA | LN833581.1 | - | 11 - 12 | 1714 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
