Assay Details
Gene Symbol: AMH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001317263.1 | NP_001304192.1 | 3 - 4 | 702 | 64 | |
GenBank mRNA | JF330269.1 | - | 3 - 4 | 702 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
