Assay Details
Gene Symbol: SEL1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001317264.1 | NP_001304193.1 | 16 - 17 | 1694 | 65 | |
XM_023627707.1 | XP_023483475.1 | 1612 | 65 | |||
GenBank mRNA | JX846917.1 | - | 16 - 17 | 1694 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
