Assay Details
Gene Symbol: KRT23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001346192.1 | NP_001333121.1 | 2 - 3 | 1488 | 81 | |
XM_023651631.1 | XP_023507399.1 | 614 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
