Assay Details
Gene Symbol: KRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001346204.2 | NP_001333133.2 | 5 - 6 | 1257 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001346204.2 | NP_001333133.2 | 5 - 6 | 1257 | 70 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |