Assay Details
Gene Symbol: BABAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502017.5 | XP_001502067.1 | 1008 | 79 | ||
XM_023619317.1 | XP_023475085.1 | 899 | 79 | |||
GenBank mRNA | JL616309.1 | - | 9 - 10 | 937 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
